Potassium perchlorate, anhydrous 99.0-100.5% ACS

Supplier: BeanTown Chemical

218935-100G 218935-500G
BT218935-100GEA 41.33 USD
BT218935-100G BT218935-500G
Potassium perchlorate, anhydrous 99.0-100.5% ACS
Potassium perchlorate

CAS: 7778-74-7; EC No: 231-912-9; MDL No: MFCD00011362; RTECS: SC9700000 UN No: UN1489; Haz Class: 5.1; Packing Group: II -100 Mesh Powder; Linear Formula: KClO4; MW: 138.55 Melting Point: 400° (decomposes) Density (g/mL): 2.52 Hygroscopic


  • RTECS: SC9700000
  • Hygroscopic
  • -100 Mesh Powder

Formula: ClKO₄
MW: 138.55 g/mol
Melting Pt: 400 °C (dec.)
Density: 2.52 g/mL
MDL Number: MFCD00011362
CAS Number: 7778-74-7
EINECS: 231-912-9
UN: 1489
ADR: 5.1,II

Order Now

Specification Test Results

Assay 99.0-100.5%
pH of 5% slurry 5.0-7.0
Insoluble matter ≤0.005%
Chloride (Cl) ≤0.003%
Sulfate (SO4) ≤0.001%
Heavy metals (as Pb) ≤0.0005%
Calcium (Ca) ≤0.005%
Iron (Fe) ≤0.0005%
Sodium (Na) ≤0.02%
Mesh size 95-100%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR