Sodium nitrite ≥98%
Supplier: BeanTown Chemical
|
|
Synonyms:
Diazotizing salts
218830-1KG
218830-5KG
BT218830-1KGEA
80.25
USD
BT218830-1KG
BT218830-5KG
Sodium nitrite ≥98%
Sodium nitrite
CAS: 7632-00-0; EC No: 231-555-9; MDL No: MFCD00011118; RTECS: RA1225000
UN No: UN1500; Haz Class: 5.1 (6.1); Packing Group: III
Solid; Linear Formula: NaNO2; MW: 69.00
Melting Point: 271°; Boiling Point: 320°
Density (g/mL): 2.168
Hygroscopic
- RTECS: RA1225000
- Hygroscopic
Formula:
NaNO₂ MW: 69 g/mol Boiling Pt: 320 Melting Pt: 284 Density: 2.168 |
MDL Number:
MFCD00011118 CAS Number: 7632-00-0 UN: 1500 ADR: 5.1 (6.1) III Merck Index: 14,8648 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...