2-Bromo-5-fluoropyridine ≥98%

Supplier: BeanTown Chemical

218470-1G 218470-5G
BT218470-1GEA 38 USD
BT218470-1G BT218470-5G
2-Bromo-5-fluoropyridine ≥98%
2-Bromo-5-fluoropyridine

CAS: 41404-58-4; MDL No: MFCD00234011 UN No: UN2811; Haz Class: 6.1; Packing Group: III Solid; Molecular Formula: C5H3BrFN; MW: 175.99 Melting Point: 30-31°; Boiling Point: 80-83°/44 mmHg; Flash point: 73°C (163°F)


  • Solid

Formula: C₅H₃BrFN
MW: 175.99 g/mol
Boiling Pt: 80…83 °C/44 mmHg
Melting Pt: 30…31 °C
Flash Pt: 73 °C (163 °F)
MDL Number: MFCD00234011
CAS Number: 41404-58-4
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR