Iodic acid ≥99.5% ACS
Supplier: BeanTown Chemical
218395-250G
218395-50G
BT218395-250GEA
262
USD
BT218395-250G
BT218395-50G
Iodic acid ≥99.5% ACS
Iodic acid
CAS: 7782-68-5; EC No: 231-962-1; MDL No: MFCD00011348
UN No: UN3085; Haz Class: 5.1 (8); Packing Group: II
Powder/Chunks; Molecular Formula: HIO3; MW: 175.91
Light Sensitive
- Light Sensitive
- Powder/Chunks
Formula:
HIO₃ MW: 175.91 g/mol Melting Pt: 110 |
MDL Number:
MFCD00011348 CAS Number: 7782-68-5 EINECS: 231-962-1 UN: 3085 ADR: 5.1 II Merck Index: 13,05034 |
Specification Test Results
Assay | 99.5% min. |
Insoluble matter | ≤0.01% |
Residue after ignition | ≤0.02% |
Chloride & bromide | ≤0.02% |
Sulfate (SO4) | ≤0.015% |
Heavy metals (as Pb) | ≤0.001% |
Iron (Fe) | ≤0.002% |
Iodide (I) | ≤0.01% |
Nitrogen compounds (as N) | ≤0.1% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...