Calcium hydroxide ≥95.0% ACS
Supplier: BeanTown Chemical
Synonyms:
Slaked lime
218185-1KG
218185-250G
BT218185-1KGEA
83.73
USD
BT218185-1KG
BT218185-250G
Calcium hydroxide ≥95.0% ACS
Calcium hydroxide
CAS: 1305-62-0; EC No: 215-137-3; MDL No: MFCD00010901; RTECS: EW2800000
UN No: UN3262; Haz Class: 8; Packing Group: III
Powder; Linear Formula: Ca(OH)2; MW: 74.09
Density (g/mL): 2.24
Air Sensitive
- RTECS: EW2800000
- Air Sensitive
- Powder
Formula:
Ca(OH)₂ MW: 74.09 g/mol Melting Pt: 550 Density: 2.24 g/mL |
MDL Number:
MFCD00010901 CAS Number: 1305-62-0 EINECS: 215-137-3 UN: 3262 ADR: 8 III Merck Index: 13,01675 |
Specification Test Results
Assay | 95.0% min. |
Calcium carbonate | ≤3.0% |
Insoluble in HCl | ≤0.03% |
Chloride (Cl) | ≤0.03% |
Sulfur compounds (as SO4) | ≤0.1% |
Heavy metals (as Pb) | ≤0.003% |
Iron (Fe) | ≤0.05% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...