Silver nitrate 1.0 N in water volumetric solution

Supplier: BeanTown Chemical

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

218085-1L 218085-500ML
BT218085-1LEA 812 USD
BT218085-1L BT218085-500ML
Silver nitrate 1.0 N in water volumetric solution
Silver nitrate

CAS: 7761-88-8; EC No: 231-853-9; MDL No: MFCD00003414; RTECS: VW4725000 UN No: UN3264; Haz Class: 8; Packing Group: II Liquid; Molecular Formula: AgNO3; MW: 169.87 Density (g/mL): 1.135 Light Sensitive


  • RTECS: VW4725000
  • Light Sensitive
  • Liquid

Formula: AgNO₃
Density: 1.135 g/mL
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR