Formic acid ≥96% ACS
Supplier: BeanTown Chemical
Synonyms:
Aminic Acid, Methanoic acid
218040-2KG
218040-500G
BT218040-2KGEA
181.74
USD
BT218040-2KG
BT218040-500G
Formic acid ≥96% ACS
Formic acid
CAS: 64-18-6; EC No: 200-579-1; MDL No: MFCD00003297; RTECS: LQ4900000
UN No: UN1779; Haz Class: 8 (3); Packing Group: II
Liquid; Linear Formula: HCOOH; MW: 46.03
Melting Point: 8°; Boiling Point: 100-101°; Flash point: 48°C (119°F)
Density (g/mL): 1.22; Refractive Index: 1.371
- RTECS: LQ4900000
- Refractive index: 1.371
- Liquid
Formula:
HCO₂H MW: 46.03 Boiling Pt: 100…101 °C Melting Pt: 8 °C Density: 1.22 g/mL Flash Pt: 48 °C (119 °F) |
MDL Number:
MFCD00003297 CAS Number: 64-18-6 EINECS: 200-579-1 UN: 1779 ADR: 8 II |
|
Specification Test Results
Assay | 96.0% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...