Hydrochloric acid 36.5 - 38.0% ACS
Supplier: BeanTown Chemical
217905-6LB
217905-6X6LB
BT217905-6LBEA
65
USD
BT217905-6LB
BT217905-6X6LB
Hydrochloric acid 36.5 - 38.0% ACS
Hydrochloric acid
CAS: 7647-01-0; EC No: 231-595-7; MDL No: MFCD00011324; RTECS: MW4025000
UN No: UN1789; Haz Class: 8; Packing Group: II
Liquid; Molecular Formula: HCl; MW: 36.46
Boiling Point:
- RTECS: MW4025000
- Liquid
Formula:
HCl Boiling Pt: >100 °C Density: 1.19 g/mL |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,II |
Specification Test Results
Assay | 36.5-38.0% |
Color (APHA) | ≤10 |
Extractable organic substances | ≤5ppm |
Arsenic (As) | ≤0.01ppm |
Ammonium (NH4) | ≤3ppm |
Bromide (Br) | ≤0.005% |
Free chloride (Cl) | ≤1ppm |
Heavy metals (as Pb) | ≤1ppm |
Iron (Fe) | ≤0.2ppm |
Residue after ignition | ≤5ppm |
Sulfate (SO4) | ≤1ppm |
Sulfite (SO3) | ≤1ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...