1,1'-Oxybis(3,3,3-trifluoro-1-propanol)hemihydrate ≥96%
Supplier: BeanTown Chemical
217735-1G
217735-5G
BT217735-1GEA
125
USD
BT217735-1G
BT217735-5G
1,1'-Oxybis(3,3,3-trifluoro-1-propanol)hemihydrate ≥96%
1,1'-Oxybis(3,3,3-trifluoro-1-propanol)hemihydrate
CAS: 936107-87-8; MDL No: MFCD00156074
UN No: UN1988; Haz Class: 3 (6.1); Packing Group: III
Liquid; Molecular Formula: C6H8F6O3·0.5H2O ; MW: 251.12
- Liquid
Formula:
C₆H₈F₆O₃·½H₂O MW: 251.12 g/mol (242.12 anhy) |
MDL Number:
MFCD00156074 CAS Number: 936107-87-8 ADR: 3 (6.1) III |
Specification Test Results
Assay | 96% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...