Potassium hydroxide 0.5 N water Volumetric

Supplier: BeanTown Chemical

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

217720-1L 217720-20L 217720-4L
BT217720-1LEA 31.95 USD
BT217720-1L BT217720-20L BT217720-4L
Potassium hydroxide 0.5 N water Volumetric
Potassium hydroxide

CAS: 1310-58-3; EC No: 215-181-3; MDL No: MFCD00003553 UN No: UN1814; Haz Class: 8; Packing Group: III Liquid; Linear Formula: KOH; MW: 56.11 Density (g/mL): 1.023


  • Liquid

Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Density: 1.023 g/mL
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8 III
Merck Index: 12,07806

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR