Hexamethyldisiloxane ≥99.5%

Supplier: BeanTown Chemical

Synonyms: Silicone fluid, HMDSO

217705-100G 217705-25G
BT217705-100GEA 69.2 USD
BT217705-100G BT217705-25G
Hexamethyldisiloxane ≥99.5%
Hexamethyldisiloxane

CAS: 107-46-0; EC No: 203-492-7; MDL No: MFCD00008265; RTECS: JM9237000 UN No: UN1993; Haz Class: 3; Packing Group: II Liquid; Linear Formula: (CH3)3SiOSi(CH3)3; Molecular Formula: C6H18OSi2; MW: 162.38 Melting Point: -59°; Boiling Point: 101°; Flash point: -2°C (28°F) Density (g/mL): 0.764; Refractive Index: 1.377


  • RTECS: JM9237000
  • Refractive index: 1.377
  • Liquid

Formula: C₆H₁₈OSi₂
MW: 162.38 g/mol
Boiling Pt: 101
Melting Pt: -59
Density: 0.764 g/mL
Flash Pt: –2 °C (28 °F)
MDL Number: MFCD00008265
CAS Number: 107-46-0
EINECS: 203-492-7
UN: 1993
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03d0d18c44dfa536ffcc853891b6f469333a38c5&vid=4959fefab2528efb6003e1f23c57966b65984db6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR