Xanthone ≥99%
Supplier: BeanTown Chemical
Synonyms:
Xanthen-9-one, 9H-Xanthen-9-one
217425-100G
217425-25G
BT217425-100GEA
95
USD
BT217425-100G
BT217425-25G
Xanthone ≥99%
Xanthone
CAS: 90-47-1; EC No: 201-997-7; MDL No: MFCD00005060; RTECS: ZD5711000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Powder; Molecular Formula: C13H8O2; MW: 196.21
Melting Point: 174-178°; Boiling Point: 350-351°
- RTECS: ZD5711000
- Powder
Formula:
C₁₃H₈O₂ MW: 196.21 g/mol Boiling Pt: 349 Melting Pt: 174 |
MDL Number:
MFCD00005060 CAS Number: 90-47-1 EINECS: 201-997-7 ADR: 6.1 III Merck Index: 12,10196 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...