n-Pentane 99,0 HPLC

Supplier: BeanTown Chemical

Synonyms: Pentane

217360-1L 217360-4L 217360-4X1L 217360-4X4L
BT217360-1LEA 88.04 USD
BT217360-1L BT217360-4L BT217360-4X1L BT217360-4X4L
n-Pentane 99,0 HPLC
n-Pentane

CAS: 109-66-0; EC No: 203-692-4; MDL No: MFCD00009498; RTECS: RZ9450000 UN No: UN1265; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3(CH2)3CH3; Molecular Formula: C5H12; MW: 72.15 Melting Point: -130°; Boiling Point: 36°; Flash point: -49°C (-56°F) Density (g/mL): 0.626; Refractive Index: 1.358


  • RTECS: RZ9450000
  • Refractive index: 1.358
  • Liquid

Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 36 °C
Melting Pt: -130
Density: 0.626 g/mL
Flash Pt: –49 °C (–56 °F)
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3 II
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR