Ethanol denatured
Supplier: BeanTown Chemical
|
|
CAS: 64-17-5; EC No: 200-578-6; MDL No: MFCD00003568; RTECS: KQ6300000
UN No: UN1987; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3CH2OH; Molecular Formula: C2H6O; MW: 46.08
Melting Point: -90°; Boiling Point: 78°; Flash point: 11°C (52°F)
Density (g/mL): 0.789
Hygroscopic
- RTECS: KQ6300000
- Hygroscopic
Formula:
H₃CCH₂OH Boiling Pt: 78 °C Melting Pt: –90 °C Density: 0.789 g/mL Flash Pt: 11 °C (52 °F) |
MDL Number:
MFCD00003568 CAS Number: 64-17-5 EINECS: 200-578-6 UN: 1987 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...