Tetrabutylammonium tribromide ≥98%

Supplier: BeanTown Chemical

Synonyms: Tetra-n-butylammonium tribromide, N,N,N,N-Tetrabutylammonium tribromide

217080-100G 217080-25G
BT217080-100GEA 141 USD
BT217080-100G BT217080-25G
Tetrabutylammonium tribromide ≥98%
Tetrabutylammonium tribromide

CAS: 38932-80-8; MDL No: MFCD00012110 UN No: UN3261; Haz Class: 8; Packing Group: III Crystalline; Linear Formula: [CH3(CH2)3]4N(Br3); Molecular Formula: C16H36Br3N; MW: 482.18 Melting Point: 71-76°


  • Crystalline

Formula: C₁₆H₃₆Br₃N
Melting Pt: 71…76 °C
MDL Number: MFCD00012110
CAS Number: 38932-80-8
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR