(1S)-(+)-Camphor-10-sulfonyl chloride ≥97%
Supplier: BeanTown Chemical
CAS: 21286-54-4; MDL No: MFCD00064156
UN No: UN3261; Haz Class: 8; Packing Group: II
Crystalline; Molecular Formula: C10H15ClO3S ; MW: 250.74
Melting Point: 65-67°
Optical Rotation: [α]22/D +33°, c = 1 in chloroform
Moisture Sensitive
- Optical rotation: [α]22/D +33°, c = 1 in chloroform
- Moisture Sensitive
- Crystalline
Formula:
C₁₀H₁₅ClO₃S Melting Pt: 65…67 °C |
MDL Number:
MFCD00064156 CAS Number: 21286-54-4 UN: 3261 ADR: 8,II |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...