Graphite (synthetic) ≥99.9995% (trace metals basis), powder -325 mesh
Supplier: BeanTown Chemical
216880-113G
216880-227G
216880-28G
BT216880-113GEA
182.57
USD
BT216880-113G
BT216880-227G
BT216880-28G
Graphite (synthetic) ≥99.9995% (trace metals basis), powder -325 mesh
Graphite
CAS: 7782-42-5; EC No: 231-955-3; MDL No: MFCD00144065; RTECS: MD9659600
UN No: UN1325; Haz Class: 4.1; Packing Group: III
Powder, UCP-2 grade, Ultra "F" purity; Molecular Formula: C; MW: 12.011
- RTECS: MD9659600
- Powder, UCP-2 grade, Ultra "F" purity
Formula:
C MW: 12.01 g/mol Melting Pt: 3827 |
MDL Number:
MFCD00144065 CAS Number: 7782-42-5 EINECS: 231-955-3 ADR: 4.1 III Merck Index: 12,04560 |
Specification Test Results
Assay | 99.9995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...