2-Methylbenzylamine ≥98%

Supplier: BeanTown Chemical

216840-1G 216840-25G 216840-5G
BT216840-1GEA 28 USD
BT216840-1G BT216840-25G BT216840-5G
2-Methylbenzylamine ≥98%
2-Methylbenzylamine

CAS: 89-93-0; EC No: 201-952-1; MDL No: MFCD00008112; RTECS: DP5800000 UN No: UN2735; Haz Class: 8; Packing Group: III Liquid; Molecular Formula: C8H11N; MW: 121.18 Boiling Point: 199°; Flash point: 84°C (183°F) Density (g/mL): 0.977; Refractive Index: 1.544 Air Sensitive


  • RTECS: DP5800000
  • Refractive index: 1.544
  • Air Sensitive
  • Liquid

Formula: C₈H₁₁N
Boiling Pt: 199 °C
Density: 0.977 g/mL
Flash Pt: 84 °C (183 °F)
MDL Number: MFCD00008112
CAS Number: 89-93-0
EINECS: 201-952-1
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR