Periodic acid ≥99%
Supplier: BeanTown Chemical
Synonyms:
Orthoperiodic acid
, ortho-Periodic acid
216725-1KG
216725-250G
216725-50G
BT216725-1KGEA
602.56
USD
BT216725-1KG
BT216725-50G
BT216725-250G
Periodic acid ≥99%
Periodic acid
- Hygroscopic, Light Sensitive
- Crystalline
Formula:
H₅IO₆ MW: 227.94 g/mol Melting Pt: 122 °C |
MDL Number:
MFCD00011344 CAS Number: 10450-60-9 EINECS: 233-937-0 UN: 3085 ADR: 5.1,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...