Quinoline ≥98%
Supplier: BeanTown Chemical
216465-1KG
216465-250G
216465-50G
BT216465-1KGEA
199.33
USD
BT216465-1KG
BT216465-250G
BT216465-50G
Quinoline ≥98%
Quinoline
CAS: 91-22-5; EC No: 202-051-6; MDL No: MFCD00006736; RTECS: VA9275000
UN No: UN2656 ; Haz Class: 6.1; Packing Group: III
Liquid; Molecular Formula: C9H7N; MW: 129.16
Melting Point: -17- -13°; Boiling Point: 237°; Flash point: 101°C (214°F)
Density (g/mL): 1.093; Refractive Index: 1.625
Hygroscopic , Light Sensitive
- RTECS: VA9275000
- Refractive index: 1.625
- Hygroscopic, Light Sensitive
- Liquid
Formula:
C₉H₇N Boiling Pt: 237 °C Melting Pt: –17– –13 °C Density: 1.093 g/mL Flash Pt: 101 °C (214 °F) |
MDL Number:
MFCD00006736 CAS Number: 91-22-5 EINECS: 202-051-6 UN: 2656 ADR: 6.1,III |
|
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...