Benzalkonium chloride 50% (w/w) in water

Supplier: BeanTown Chemical

Synonyms: Quaternary ammonium compounds, benzyl-C8-18-alkyldimethyl, chlorides, Alkylbenzyldimethylammonium chloride

216360-2.5L 216360-500ML
BT216360-2.5LEA 227.43 USD
BT216360-2.5L BT216360-500ML
Benzalkonium chloride 50% (w/w) in water
Benzalkonium chloride

CAS: 63449-41-2; EC No: 264-151-6; MDL No: MFCD00145757; RTECS: BO3151000 UN No: UN3265; Haz Class: 8; Packing Group: III Liquid; Linear Formula: C6H5CH2N(CH3)2RCl, R = C8H17 to C18H37 Density (g/mL): 0.99


  • RTECS: BO3151000
  • Liquid

Formula: C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
Density: 0.99 g/mL
MDL Number: MFCD00145757
CAS Number: 63449-41-2
EINECS: 264-151-6
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR