2,2,2-Trifluoroethanol 99
Supplier: BeanTown Chemical
Synonyms:
Trifluoro Ethanol, TFE
215880-100G
215880-500G
BT215880-100GEA
48.98
USD
BT215880-100G
BT215880-500G
2,2,2-Trifluoroethanol 99
2,2,2-Trifluoroethanol
CAS: 75-89-8; EC No: 200-913-6; MDL No: MFCD00004672; RTECS: KM5250000
UN No: UN1992; Haz Class: 3 (6.1); Packing Group: III
Liquid; Linear Formula: CF3CH2OH; MW: 100.04
Melting Point: -44°; Boiling Point: 77-80°; Flash point: 29°C (84°F)
Density (g/mL): 1.373; Refractive Index: 1.300
- RTECS: KM5250000
- Refractive index: 1.300
- Liquid
Formula:
C₂H₃F₃O MW: 100.04 g/mol Boiling Pt: 74.1 Melting Pt: -43.5 Density: 1.373 g/mL Flash Pt: 29 °C (84 °F) |
MDL Number:
MFCD00004672 CAS Number: 75-89-8 EINECS: 200-913-6 UN: 1992 ADR: 6.1 III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...