Acrylic acid ≥99% stabilized
Supplier: BeanTown Chemical
Synonyms:
Prop-2-enoic acid
215770-500G
215770-100G
215770-2.5KG
BT215770-100GEA
42.73
USD
BT215770-100G
BT215770-500G
BT215770-2.5KG
Acrylic acid ≥99% stabilized
Acrylic acid
- RTECS: AS4375000
- Refractive index: 1.420
- Liquid
Formula:
C₃H₄O₂ MW: 72.06 g/mol Boiling Pt: 139 °C Melting Pt: 13 °C Density: 1.051 g/mL Flash Pt: 46 °C (115 °F) |
MDL Number:
MFCD00004367 CAS Number: 79-10-7 EINECS: 201-177-9 UN: 2218 ADR: 8,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...