Allylthiourea ≥98%
Supplier: BeanTown Chemical
Synonyms:
N-Allylthiourea, Tiosinamine, 1-Allyl-2-thiourea, Allyl thiourea, Rhodalline, Thiosinamine, N-Allylthiourea
215510-100G
215510-25G
BT215510-100GEA
78.78
USD
BT215510-100G
BT215510-25G
Allylthiourea ≥98%
Allylthiourea
CAS: 109-57-9; EC No: 203-683-5; MDL No: MFCD00004940; RTECS: YR8050000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Crystalline/Powder; Linear Formula: H2=CHCH2NHCSNH2; Molecular Formula: C4H8N2S; MW: 116.19
Melting Point: 70-72°
Density (g/mL): 1.11
- RTECS: YR8050000
- Crystalline/Powder
Formula:
H₂CCHCH₂NHC(S)NH₂ MW: 116.19 g/mol Boiling Pt: 191 Melting Pt: 74 Density: 1.11 g/mL |
MDL Number:
MFCD00004940 CAS Number: 109-57-9 EINECS: 203-683-5 UN: 2811 ADR: 6.1 III Merck Index: 12,09500 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...