Zinc chloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
Zinc dichloride, Butter of zinc, Dichlorozinc, Zinc (II) chloride
215405-100G
215405-2.5KG
215405-500G
BT215405-100GEA
31.29
USD
BT215405-100G
BT215405-2.5KG
BT215405-500G
Zinc chloride ≥98%
Zinc chloride
CAS: 7646-85-7; EC No: 231-592-0; MDL No: MFCD00011295; RTECS: ZH1400000
UN No: UN2331; Haz Class: 8; Packing Group: III
Powder; Linear Formula: ZnCl2; MW: 136.29
Melting Point: 293°; Boiling Point: 732°
Density (g/mL): 2.907
Hygroscopic
- RTECS: ZH1400000
- Hygroscopic
- Powder
Formula:
ZnCl₂ MW: 136.3 g/mol Boiling Pt: 732 Melting Pt: 283 Density: 2.907 g/mL |
MDL Number:
MFCD00011295 CAS Number: 7646-85-7 EINECS: 231-592-0 UN: 2331 ADR: 8 III Merck Index: 13,10185 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...