Cetrimonium chloride ≥96%

Supplier: BeanTown Chemical
Danger

215345-100G 215345-25G 215345-5G
BT215345-100GEA 274.61 USD
BT215345-100G BT215345-25G BT215345-5G
Cetrimonium chloride ≥96%
Cetrimonium chloride

CAS: 112-02-7; EC No: 203-928-6; MDL No: MFCD00011773; RTECS: ML9145000 UN No: UN1759; Haz Class: 8; Packing Group: III Powder; Linear Formula: CH3(CH2)15N(Cl)(CH3)3; Molecular Formula: C19H42ClN; MW: 320.00 Melting Point: 232-237° Density (g/mL): 0.968; Refractive Index: 1.378


  • RTECS: ML9145000
  • Refractive index: 1.378
  • Powder

Formula: C₁₉H₄₂ClN
Melting Pt: 232…237 °C
Density: 0.968 g/mL
MDL Number: MFCD00011773
CAS Number: 112-02-7
EINECS: 203-928-6
UN: 1759
ADR: 8,III

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR