1,2,4-Trichlorobenzene 99 Spectrophotometry

Supplier: BeanTown Chemical

215305-1L 215305-250ML 215305-4L
BT215305-1LEA 73 USD
BT215305-1L BT215305-250ML BT215305-4L
1,2,4-Trichlorobenzene 99 Spectrophotometry
1,2,4-Trichlorobenzene

CAS: 120-82-1; EC No: 204-428-0; MDL No: MFCD00000547 UN No: UN2321; Haz Class: 6.1; Packing Group: III Liquid; Molecular Formula: C6H3Cl3; MW: 181.45 Melting Point: 16°; Boiling Point: 214°; Flash point: 110°C (230°F) Density (g/mL): 1.454; Refractive Index: 1.572


  • Refractive index: 1.572
  • Liquid

Formula: C₆H₃Cl₃
MW: 181.45 g/mol
Boiling Pt: 214.4
Melting Pt: 17
Density: 1.454 g/mL
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00000547
CAS Number: 120-82-1
EINECS: 204-428-0
UN: 2321
ADR: 6.1 III
Merck Index: 13,09704

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR