1-Methylimidazole (NMI) ≥99%
Supplier: BeanTown Chemical
Synonyms:
N-Methylimidazole, 1-Methyl-1H-imidazole, N-Methyl-1H-imidazole, NMI
214950-100G
214950-2KG
214950-500G
BT214950-100GEA
37.48
USD
BT214950-100G
BT214950-2KG
BT214950-500G
1-Methylimidazole (NMI) ≥99%
1-Methylimidazole
CAS: 616-47-7; EC No: 210-484-7; MDL No: MFCD00005292; RTECS: NI7000000
UN No: UN2922; Haz Class: 8 (6.1); Packing Group: III
Liquid; Molecular Formula: C4H6N2; MW: 82.10
Melting Point: -6°; Boiling Point: 198°; Flash point: 92°C (198°F)
Density (g/mL): 1.03; Refractive Index: 1.495
Hygroscopic
- RTECS: NI7000000
- Refractive index: 1.495
- Hygroscopic
- Liquid
Formula:
C₄H₆N₂ MW: 82.11 g/mol Boiling Pt: 198 °C Melting Pt: –6 °C Density: 1.03 g/mL Flash Pt: 92 °C (198 °F) |
MDL Number:
MFCD00005292 CAS Number: 616-47-7 EINECS: 210-484-7 UN: 2922 ADR: 8 III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...