Sodium periodate 99.8-100.3% ACS

Supplier: BeanTown Chemical

Synonyms: Sodium metaperiodate

214445-2KG 214445-100G 214445-500G
BT214445-100GEA 128.89 USD
BT214445-100G BT214445-2KG BT214445-500G
Sodium periodate 99.8-100.3% ACS
Sodium periodate

  • RTECS: SD4550000
  • Hygroscopic, Light Sensitive
  • Crystalline/Powder

Formula: NaIO₄
MW: 213.89 g/mol
Melting Pt: 300 °C
Density: 3.86 g/mL
MDL Number: MFCD00003534
CAS Number: 7790-28-5
EINECS: 232-197-6
UN: 1479
ADR: 5.1,II

Order Now

Specification Test Results

Assay 99.8-100.3%
Loss on drying ≤0.5%
Aqueous solubility, in 100ml at 50°C 37g
pH 4.0-5.0
Other halogens (as Cl) ≤0.02%
Manganese (Mn) ≤3ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR