1-Propanol 99,0 HPLC
Supplier: BeanTown Chemical
Synonyms:
Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane
213970-1L
213970-4L
213970-4X1L
BT213970-1LEA
70.32
USD
BT213970-1L
BT213970-4L
BT213970-4X1L
1-Propanol 99,0 HPLC
1-Propanol
CAS: 71-23-8; EC No: 200-746-9; MDL No: MFCD00002941; RTECS: UH8225000
UN No: UN1274; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3CH2CH2OH; Molecular Formula: C3H8; MW: 60.10
Melting Point: -127°; Boiling Point: 97°; Flash point: 22°C (72°F)
Density (g/mL): 0.804; Refractive Index: 1.384
- RTECS: UH8225000
- Refractive index: 1.384
- Liquid
Formula:
H₃CCH₂CH₂OH MW: 60.1 g/mol Boiling Pt: 97 Melting Pt: -127 Density: 0.804 g/mL Flash Pt: 22 °C (72 °F) |
MDL Number:
MFCD00002941 CAS Number: 71-23-8 EINECS: 200-746-9 UN: 1274 ADR: 3 II Merck Index: 13,07934 |
|
Specification Test Results
Assay | 99.0% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...