Bismuth(III) nitrate oxide ≥99.9%, Technical Grade

Supplier: BeanTown Chemical

Synonyms: Bismuth (III) oxide nitrate, (Nitrooxy)oxobismuthine, Bismuth (III) oxynitrate

213600-10G 213600-250G 213600-50G
BT213600-10GEA 39 USD
BT213600-10G BT213600-250G BT213600-50G
Bismuth(III) nitrate oxide ≥99.9%, Technical Grade
Bismuth (III) nitrate oxide

CAS: 10361-46-3; EC No: 233-792-3; MDL No: MFCD00064844 UN No: UN1477; Haz Class: 5.1; Packing Group: II Powder; Linear Formula: BiONO3; MW: 286.98


  • Powder

Formula: BiNO₄
MW: 286.98
Density: 4.93
MDL Number: MFCD00064844
CAS Number: 10361-46-3
EINECS: 233-792-3
UN: 1477
ADR: 5.1 II

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR