Methylenediphosphonic acid ≥97%
Supplier: BeanTown Chemical
CAS: 1984-15-2; EC No: 217-851-0; MDL No: MFCD00002139; RTECS: SZ9138000
UN No: UN3261; Haz Class: 8; Packing Group: III
Crystalline; Linear Formula: CH2[P(O)(OH)2]2; Molecular Formula: CH6O6P2; MW: 176.00
Melting Point: 197-199°
- RTECS: SZ9138000
- Crystalline
Formula:
CH₆O₆P₂ Melting Pt: 197…199 °C |
MDL Number:
MFCD00002139 CAS Number: 1984-15-2 EINECS: 217-851-0 UN: 3261 ADR: 8,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...