Arsenic (V) oxide ≥99.9%

Supplier: BeanTown Chemical

Synonyms: Arsenic oxide, Arsenic pentoxide, di-Arsenic pentaoxide

213285-100G 213285-25G
BT213285-100GEA 424 USD
BT213285-100G BT213285-25G
Arsenic (V) oxide ≥99.9%
Arsenic (V) oxide

CAS: 1303-28-2; EC No: 215-116-9; MDL No: MFCD00003434; RTECS: CG2275000 UN No: UN1559; Haz Class: 6.1; Packing Group: II -100 Mesh Powder; Molecular Formula: As2O5; MW: 229.79 Melting Point: 315° (decomposes) Hygroscopic


  • RTECS: CG2275000
  • Hygroscopic
  • -100 Mesh Powder

Formula: As₂O₅
MW: 229.84 g/mol
Melting Pt: 730 (dec.)
MDL Number: MFCD00003434
CAS Number: 1303-28-2
EINECS: 215-116-9
UN: 1559
ADR: 6.1 II
Merck Index: 12,00839

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR