Hexamethyldisilathiane ≥98%
Supplier: BeanTown Chemical
CAS: 3385-94-2; EC No: 222-201-4; MDL No: MFCD00014851
UN No: UN1993; Haz Class: 3; Packing Group: II
Liquid, packaged in ampules; Linear Formula: (CH3)3SiSSi(CH3)3; Molecular Formula: C9H18SSi2; MW: 178.44
Boiling Point: 164°; Flash point: 23°C (73°F)
Density (g/mL): 0.851; Refractive Index: 1.460
- Refractive index: 1.460
- Liquid, packaged in ampules
Formula:
C₆H₁₈SSi₂ Boiling Pt: 164 °C Density: 0.851 g/mL Flash Pt: 23 °C (73 °F) |
MDL Number:
MFCD00014851 CAS Number: 3385-94-2 EINECS: 222-201-4 ADR: 3,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...