Acrylic acid, low water content ≥99.5% stabilised
Supplier: BeanTown Chemical
Synonyms:
Prop-2-enoic acid
213040-2.5L
213040-500ML
BT213040-2.5LEA
127.66
USD
BT213040-2.5L
BT213040-500ML
Acrylic acid, low water content ≥99.5% stabilised
Acrylic acid
CAS: 79-10-7; EC No: 201-177-9; MDL No: MFCD00004367; RTECS: AS4375000
UN No: UN2218; Haz Class: 8 (3); Packing Group: II
Liquid; Linear Formula: H2C=CHCO2H ; MW: 72.06
Melting Point: 13°; Boiling Point: 139°; Flash point: 46°C (115°F)
Density (g/mL): 1.051
- RTECS: AS4375000
- Liquid
Formula:
C₃H₄O₂ Boiling Pt: 139 °C Melting Pt: 13 °C Density: 1.051 g/mL Flash Pt: 46 °C (115 °F) |
MDL Number:
MFCD00004367 CAS Number: 79-10-7 EINECS: 201-177-9 UN: 2218 ADR: 8,II |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...