Chloramine-T (sodium salt) trihydrate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Tosylchloramide sodium trihydrate, Chlorine bleaching solution, Chloramine T trihydrate, Chloramine T sodium salt trihydrate, N-Chloro-4-toluenesulfonamide sodium salt, Sodium chloro(tosyl)amide trihydrate, N-Chloro-4-toluenesulphonamide sodium salt
212930-1KG
212930-250G
212930-5KG
BT212930-1KGEA
89
USD
BT212930-1KG
BT212930-250G
BT212930-5KG
Chloramine-T (sodium salt) trihydrate ≥98%
Chloramine T (sodium salt) trihydrate
CAS: 7080-50-4; EC No: 204-854-7; MDL No: MFCD00149066; RTECS: XT5616800
UN No: UN3263; Haz Class: 8; Packing Group: III
Crystalline; Molecular Formula: C7H7ClNNaO2S·3H2O; MW: 281.69
Melting Point: 167-170°; Flash point: 192°C (378°F)
Moisture Sensitive
- RTECS: XT5616800
- Moisture Sensitive
- Crystalline
Formula:
CH₃C₆H₄SO₂NClNa·3H₂O MW: 281.69 g/mol Melting Pt: 167…170 °C Flash Pt: 192 °C (378 °F) |
MDL Number:
MFCD00149066 CAS Number: 7080-50-4 EINECS: 204-854-7 UN: 3263 ADR: 8 III Merck Index: 13,02085 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...