2,3-Dimercapto-1-propanol ≥97%

Supplier: BeanTown Chemical

Synonyms: 2,3-DimercaptopropanolDithioglycerineDimercaprolDimercapto-2,3 propanol-12,3-Dimercaptopropane-1-ol2,3-Dimercaptopropan-1-olBAL

212805-5G 212805-25G 212805-1G
BT212805-5GEA 106.97 USD
BT212805-5G BT212805-25G BT212805-1G
2,3-Dimercapto-1-propanol ≥97%
2,3-Dimercapto-1-propanol

  • RTECS: UB2625000
  • Air Sensitive
  • Liquid

Formula: C₃H₈OS₂
MW: 124.22 g/mol
Boiling Pt: 120 °C/15 mmHg
Density: 1.239 g/mL
Flash Pt: 112 °C (234 °F)
MDL Number: MFCD00004864
CAS Number: 59-52-9
EINECS: 200-433-7
UN: 2810
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/81/18/2358118.jpg

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR