Bromotrimethylsilane ≥97% stabilized
Supplier: BeanTown Chemical
CAS: 2857-97-8; EC No: 220-672-0; MDL No: MFCD00000048
UN No: UN2924; Haz Class: 3(8); Packing Group: II
Liquid; Linear Formula: (CH3)3SiBr; MW: 153.09
Boiling Point: 79°; Flash point: 25°C (77°F)
Density (g/mL): 1.16; Refractive Index: 1.424
Moisture Sensitive
- Refractive index: 1.424
- Moisture Sensitive
- Liquid
Formula:
C₃H₉BrSi Boiling Pt: 79 °C Density: 1.16 g/mL Flash Pt: 25 °C (77 °F) |
MDL Number:
MFCD00000048 CAS Number: 2857-97-8 EINECS: 220-672-0 UN: 2924 ADR: 3,II |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...