Potassium hydrogen sulfate ≥97%

Supplier: BeanTown Chemical

Synonyms: Potassium bisulfate, Potassium bisulphate

212345-100G 212345-2.5KG 212345-500G
BT212345-100GEA 37.48 USD
BT212345-100G BT212345-2.5KG BT212345-500G
Potassium hydrogen sulfate ≥97%
Potassium hydrogen sulphate

CAS: 7646-93-7; EC No: 231-594-1; MDL No: MFCD00011404; RTECS: TS7200000 UN No: UN2509; Haz Class: 8; Packing Group: II Solid; Linear Formula: KHSO4; MW: 136.17 Melting Point: 195-197° Density (g/mL): 2.32 Hygroscopic


  • RTECS: TS7200000
  • Hygroscopic
  • Solid

Formula: KHSO₄
MW: 136.17 g/mol
Melting Pt: 214
Density: 2.32 g/mL
MDL Number: MFCD00011404
CAS Number: 7646-93-7
EINECS: 231-594-1
UN: 2509
ADR: 8 II
Merck Index: 13,07695

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR