Gallium arsenide ≥99.999%

Supplier: BeanTown Chemical

212275-1G 212275-5G
BT212275-1GEA 56 USD
BT212275-1G BT212275-5G
Gallium arsenide ≥99.999%
Gallium arsenide

CAS: 1303-00-0; EC No: 215-114-8; MDL No: MFCD00011017 UN No: UN1557; Haz Class: 6.1; Packing Group: II 25mm & Down Polycrystalline Pieces; Molecular Formula: GaAs; MW: 144.64 Density (g/mL): 5.31


  • 25mm & Down Polycrystalline Pieces

Formula: AsGa
MW: 144.64
Melting Pt: 1238
Density: 5.31 g/mL
MDL Number: MFCD00011017
CAS Number: 1303-00-0
EINECS: 215-114-8
UN: 1557
ADR: 6.1 II
Merck Index: 12,04365

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR