1-Bromopentane ≥99%

Supplier: BeanTown Chemical

212170-1KG 212170-250G 212170-5KG
BT212170-1KGEA 114 USD
BT212170-1KG BT212170-250G BT212170-5KG
1-Bromopentane ≥99%
1-Bromopentane

CAS: 110-53-2; EC No: 203-776-0; MDL No: MFCD00000267; RTECS: RZ9770000 UN No: UN1993; Haz Class: 3; Packing Group: III Liquid; Linear Formula: CH3(CH2)4Br; Molecular Formula: C5H11Br; MW: 151.04 Melting Point: -95°; Boiling Point: 130°; Flash point: 32°C (90°F) Density (g/mL): 1.218; Refractive Index: 1.444


  • RTECS: RZ9770000
  • Refractive index: 1.444
  • Liquid

Formula: BrH₂C(CH₂)₃CH₃
MW: 151.05 g/mol
Boiling Pt: 129
Melting Pt: -95
Density: 1.218 g/mL
Flash Pt: 32 °C (90 °F)
MDL Number: MFCD00000267
CAS Number: 110-53-2
EINECS: 203-776-0
UN: 1993
ADR: 3 III
Merck Index: 13,00606

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR