2,4,6,8-Tetramethylcyclotetrasiloxane ≥97%
Supplier: BeanTown Chemical
CAS: 2370-88-9; EC No: 219-137-4; MDL No: MFCD00039567
UN No: UN1993; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C4H16O4Si4; MW: 240.51
Melting Point: -69°; Boiling Point: 134°; Flash point: 24°C (75°F)
Density (g/mL): 0.986; Refractive Index: 1.387
Moisture Sensitive
- Refractive index: 1.387
- Moisture Sensitive
- Liquid
Formula:
C₄H₁₆O₄Si₄ Boiling Pt: 134 °C Melting Pt: –69 °C Density: 0.986 g/mL Flash Pt: 24 °C (75 °F) |
MDL Number:
MFCD00039567 CAS Number: 2370-88-9 EINECS: 219-137-4 ADR: 3,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...