Sodium methoxide ≥98%

Supplier: BeanTown Chemical

Synonyms: Sodium methylate, Sodium methanolate

212100-100G 212100-2KG 212100-500G
BT212100-100GEA 37.49 USD
BT212100-100G BT212100-2KG BT212100-500G
Sodium methoxide ≥98%
Sodium methoxide

CAS: 124-41-4; EC No: 204-699-5; MDL No: MFCD00012179; RTECS: PC3570000 UN No: UN1431; Haz Class: 4.2 (8); Packing Group: II Powder; Linear Formula: CH3ONa; MW: 54.02 Melting Point: 300°; Flash point: 33°C (91°F) Density (g/mL): 0.97 Moisture Sensitive


  • RTECS: PC3570000
  • Moisture Sensitive
  • Powder

Formula: CH₃NaO
MW: 54.02 g/mol
Melting Pt: 127
Density: 0.97 g/mL
Flash Pt: 33 °C (91 °F)
MDL Number: MFCD00012179
CAS Number: 124-41-4
EINECS: 204-699-5
UN: 1431
ADR: 4.2 II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR