4,6-Dichloro-2-methylpyrimidine ≥97%

Supplier: BeanTown Chemical

Synonyms: 2-Methyl-4,6-dichloropyrimidine

212055-1G 212055-5G
BT212055-1GEA 55 USD
BT212055-1G BT212055-5G
4,6-Dichloro-2-methylpyrimidine ≥97%
4,6-Dichloro-2-methylpyrimidine

CAS: 1780-26-3; MDL No: MFCD00090472 UN No: UN1759; Haz Class: 8; Packing Group: III Solid; Molecular Formula: C5H4Cl2N2; MW: 163.00 Melting Point: 41-46°; Flash point: 97°C (207°F)


  • Solid

Formula: C₅H₄Cl₂N₂
MW: 163.01
Melting Pt: 41…46 °C
Flash Pt: 97 °C (207 °F)
MDL Number: MFCD00090472
CAS Number: 1780-26-3
ADR: 8 III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR