Sodium peroxodisulfate ≥98%

Supplier: BeanTown Chemical

Synonyms: Sodium peroxidisulfate, Sodium peroxidisulphate, Sodium persulphate, Disodium peroxodisulphate, Peroxydisulphuric acid disodium salt, Sodium persulfate, Peroxydisulfuric acid disodium salt, Disodium peroxodisulfate

211965-1KG 211965-5KG
BT211965-1KGEA 51.85 USD
BT211965-1KG BT211965-5KG
Sodium peroxodisulfate ≥98%
Sodium peroxodisulphate

CAS: 7775-27-1; EC No: 231-892-1; MDL No: MFCD00003501; RTECS: SE0525000 UN No: UN1505; Haz Class: 5.1; Packing Group: III Crystalline; Linear Formula: Na2S2O8; MW: 238.10 Density (g/mL): 2.4


  • RTECS: SE0525000
  • Crystalline

Formula: Na₂S₂O₈
MW: 238.11 g/mol
Melting Pt: 100
Density: 2.4 g/mL
MDL Number: MFCD00003501
CAS Number: 7775-27-1
EINECS: 231-892-1
UN: 1505
ADR: 5.1 III
Merck Index: 13,08729

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR