Barium oxalate ≥99.999%

Supplier: BeanTown Chemical

211900-100G 211900-25G 211900-5G
BT211900-100GEA 333 USD
BT211900-100G BT211900-25G BT211900-5G
Barium oxalate ≥99.999%
Barium oxalate

CAS: 516-02-9; EC No: 208-216-9; MDL No: MFCD00054429 UN No: UN1564; Haz Class: 6.1; Packing Group: II Powder; Molecular Formula: C2BaO4; MW: 225.36


  • Powder

Formula: C₂BaO₄
MW: 225.35
Density: 2.658
MDL Number: MFCD00054429
CAS Number: 516-02-9
EINECS: 208-216-9
UN: 1564
ADR: 6.1 II
Merck Index: 12,01014

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR