Mercury(II) acetate 98,0 ACS

Supplier: BeanTown Chemical

Synonyms: Mercury diacetate, Mercuric acetate, Acetic acid mercury (II) salt

211855-250G 211855-50G
BT211855-250GEA 174 USD
BT211855-250G BT211855-50G
Mercury(II) acetate 98,0 ACS
Mercury (II) acetate

CAS: 1600-27-7; EC No: 216-491-1; MDL No: MFCD00012165; RTECS: AI8575000 UN No: UN1629; Haz Class: 6.1; Packing Group: II Crystalline; Linear Formula: Hg(OOCCH3)2; MW: 318.68 Melting Point: 179-182° Density (g/mL): 3.28 Light Sensitive


  • RTECS: AI8575000
  • Light Sensitive
  • Crystalline

Formula: (H₃CCOO)₂Hg
MW: 318.68 g/mol
Melting Pt: 179…182 °C
Density: 3.28 g/mL
MDL Number: MFCD00012165
CAS Number: 1600-27-7
EINECS: 216-491-1
UN: 1629
ADR: 6.1 II
Merck Index: 13,05898

Order Now

Specification Test Results

Assay 98.0% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR