(Ethyl)triphenylphosphonium bromide ≥98%

Supplier: BeanTown Chemical

211730-100G 211730-25G 211730-500G
BT211730-100GEA 89 USD
BT211730-100G BT211730-25G BT211730-500G
(Ethyl)triphenylphosphonium bromide ≥98%
(Ethyl)triphenylphosphonium bromide

CAS: 1530-32-1; EC No: 216-223-3; MDL No: MFCD00011838 UN No: UN3077; Haz Class: 9; Packing Group: III Solid; Molecular Formula: C20H20BrP ; MW: 371.26 Melting Point: 203-205° Hygroscopic


  • Hygroscopic
  • Solid

Formula: C₂₀H₂₀BrP
Melting Pt: 203…205 °C
MDL Number: MFCD00011838
CAS Number: 1530-32-1
EINECS: 216-223-3
UN: 3077
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR