Potassium hydroxide ≥85%, Technical Grade
Supplier: BeanTown Chemical
Synonyms:
Potash caustic, Potassium hydrate, Caustic potash
211720-2.5KG
211720-500G
BT211720-2.5KGEA
93.83
USD
BT211720-2.5KG
BT211720-500G
Potassium hydroxide ≥85%, Technical Grade
Potassium hydroxide
CAS: 1310-58-3; EC No: 215-181-3; MDL No: MFCD00003553; RTECS: TT2100000
UN No: UN1813; Haz Class: 8; Packing Group: II
Pellets; Linear Formula: KOH; MW: 56.11
Melting Point: 361°; Boiling Point: 1320°
Density (g/mL): 2.044
Air Sensitive , Hygroscopic
- RTECS: TT2100000
- Air Sensitive, Hygroscopic
- Pellets
Formula:
KOH MW: 56.11 g/mol Boiling Pt: 1320 Melting Pt: 360 Density: 2.044 g/mL |
MDL Number:
MFCD00003553 CAS Number: 1310-58-3 EINECS: 215-181-3 UN: 1813 ADR: 8 II Merck Index: 12,07806 |
Specification Test Results
Assay | 85% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...