Ethyl chloroformate ≥97%
Supplier: BeanTown Chemical
Synonyms:
Chloroformic acid ethyl ester
211460-100G
211460-500G
BT211460-100GEA
86.46
USD
BT211460-100G
BT211460-500G
Ethyl chloroformate ≥97%
Ethyl chloroformate
CAS: 541-41-3; EC No: 208-778-5; MDL No: MFCD00000644; RTECS: LQ6125000
UN No: UN1182; Haz Class: 6.1 (3,8); Packing Group: I
Liquid; Linear Formula: ClCOOCH2CH3; Molecular Formula: C3H5ClO2; MW: 108.52
Melting Point: -81°; Boiling Point: 93°; Flash point: 10°C (50°F)
Density (g/mL): 1.135; Refractive Index: 1.395
Moisture Sensitive
- RTECS: LQ6125000
- Refractive index: 1.395
- Moisture Sensitive
- Liquid
Formula:
C₃H₅ClO₂ Boiling Pt: 93 °C Melting Pt: –81 °C Density: 1.135 g/mL Flash Pt: 10 °C (50 °F) |
MDL Number:
MFCD00000644 CAS Number: 541-41-3 EINECS: 208-778-5 UN: 1182 ADR: 6.1,I |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...